Methyl stearate, 99%
Methyl stearate, 99%
Methyl stearate, 99%
Methyl stearate, 99%
Thermo Scientific Chemicals
Methyl stearate, 99%
CAS: 112-61-8 | C19H38O2 | 298.511 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA13265.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl Stearate
CAS112-61-8
Melting Point38°C to 42°C
Recommended StorageAmbient temperatures
RTECS NumberWI4460000
View more
Methyl stearate is used as a nonionic surfactant, thereby enhancing the solubility of chemicals by dissociating aggregates and unfolding proteins. It is a fatty acid ester, which is used as an emulsifier and stabilizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl stearate is used as a nonionic surfactant, thereby enhancing the solubility of chemicals by dissociating aggregates and unfolding proteins. It is a fatty acid ester, which is used as an emulsifier and stabilizer.
Solubility
Soluble in chloroform. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Cunico, L. P.; Damaceno, D. S.; Falleiro, R. M. M.; Sarup, B.; Abildskov, J.; Ceriani, R.; Gani, R. Vapour liquid equilibria of monocaprylin plus palmitic acid or methyl stearate at P=(1.20 and 2.50) kPa by using DSC technique. J. Chem. Thermodyn. 2015, 91, 108-115.
  2. Xu, S.; Zou, L.; Ling, X.; Wei, Y.; Zhang, S. Preparation and thermal reliability of methyl palmitate/methyl stearate mixture as a novel composite phase change material. Energy Build. 2014, 68, 372-375.