3-Chlorobenzaldehyde, 97%
3-Chlorobenzaldehyde, 97%
3-Chlorobenzaldehyde, 97%
3-Chlorobenzaldehyde, 97%
Thermo Scientific Chemicals
3-Chlorobenzaldehyde, 97%
CAS: 587-04-2 | C7H5ClO | 140.566 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13254.1425 g
Catalog number ALFA13254.14
Price (MYR)
311.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
311.00
EA
Add to cart
Specifications
Chemical Name or Material3-Chlorobenzaldehyde
CAS587-04-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chlorobenzaldehyde, is used as a medicine, dye intermediates and specialty chemicals. It is mainly used to produce medicine material and intermediate, chloral alcoholate and intermediate of acid mordant azurine-6B on dyestuff.
Solubility
It is soluble in alcohol, ether, and acetone. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong bases, Strong oxidizing agents, Strong reducing agents.
RUO – Research Use Only
General References:
  1. G Lu, X Li, WL Chan, ASC Chan. Titanium-catalyzed enantioselective alkynylation of aldehydes. Chemical Communications. 2002, (2), 172-173.
  2. G Lu.; YM Li.; XS Li.; ASC Chan. Synthesis and application of new chiral catalysts for asymmetric alkynylation reactions. Coordination chemistry reviews.2005, 249,(17-18), 1736-1744.