n-Octane, 98+%
n-Octane, 98+%
n-Octane, 98+%
Thermo Scientific Chemicals
n-Octane, 98+%
CAS: 111-65-9 | C8H18 | 114.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
Catalog number ALFA13181.AP
Price (MYR)
933.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
933.00
EA
Add to cart
Specifications
Chemical Name or Materialn-Octane
CAS111-65-9
Health Hazard 1H225-H304-H315-H319-H336
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
View more
N-octane is mainly used as a solvent, fuel and as one of the standard values in octane rating scale. It is also used to prevent uneven burning of fuel in internal-combustion engines. Further, it acts as an antiknock agent for high compression engine fuels, lacquer diluents, blowing agent for foam rubber. In addition, it is used in rocket propellants and in azeotropic distillations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-octane is mainly used as a solvent, fuel and as one of the standard values in octane rating scale. It is also used to prevent uneven burning of fuel in internal-combustion engines. Further, it acts as an antiknock agent for high compression engine fuels, lacquer diluents, blowing agent for foam rubber. In addition, it is used in rocket propellants and in azeotropic distillations.
Solubility
Miscible with ethyl ether, ethanol, acetone, benzene, petroleum ether and gasoline. Slightly Miscible with alcohol. Immiscible with water.
Notes
Hygroscopic. Incompatible with oxidizing agents and acids
RUO – Research Use Only
General References:
  1. Lonsdale, T. H.; Lauterbach, L.; Malca, S. H.; Nestl, B. M.; Hauer, B.; Lenz, O. H2-driven biotransformation of n-octane to 1-octanol by a recombinant Pseudomonas putida strain co-synthesizing an O2-tolerant hydrogenase and a P450 monooxygenase. Chem. Commun. 2015, 51 (90), 16173-16175.
  2. Ralphs, K.; Chansai, S.; Hardacre, C.; Burch, R.; Taylor, S. F. R.; James, S. L. Mechanochemical preparation of Ag catalysts for the n-octane-SCR de-NOx reaction: Structural and reactivity effects. Catal. Today 2015, 246, 198-206.