Tetramethylsilane, 99.9%
Tetramethylsilane, 99.9%
Tetramethylsilane, 99.9%
Tetramethylsilane, 99.9%
Thermo Scientific Chemicals
Tetramethylsilane, 99.9%
CAS: 75-76-3 | C4H12Si | 88.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA13148.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point26°C to 27°C
Chemical Name or MaterialTetramethylsilane
Melting Point-100°C
CAS75-76-3
Health Hazard 1H224
View more
Tetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
Solubility
Miscible with water, ethanol, ether and most organic solvents. Immiscible with cold sulfuric acid.
Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Internal standard for NMR spectroscopy.
  2. Kim, B. G.; Yoon, J. Y.; Yoo, C. H.; Nam, D. H.; Lee, M. H.; Seo, W. S.; Jeong, S. M. Condensation of vapor species at the outlets in high temperature chemical vapor deposition using tetramethylsilane as a precursor for SiC bulk growth. CrystEngComm 2015, 17 (16), 3148-3152.
  3. Hei, H.; Yu, S.; Shen, Y.; Li, X.; Ma, J.; Tang, B.; Tang, W. Growth of β-SiC interlayers on WC-Co substrates with varying hydrogen/tetramethylsilane flow ratio for adhesion enhancement of diamond coatings. Surf. Coat. Technol. 2015, 272, 278-284.