4-Chlorophenoxyacetic acid, 98+%
4-Chlorophenoxyacetic acid, 98+%
4-Chlorophenoxyacetic acid, 98+%
4-Chlorophenoxyacetic acid, 98+%
Thermo Scientific Chemicals
4-Chlorophenoxyacetic acid, 98+%
CAS: 122-88-3 | C8H7ClO3 | 186.591 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13065.22100 g
Catalog number ALFA13065.22
Price (MYR)
287.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
287.00
EA
Add to cart
Specifications
Chemical Name or Material4-Chlorophenoxyacetic acid
Melting Point156°C to 161°C
CAS122-88-3
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302