3-(2-Furyl)acrolein, 99%
3-(2-Furyl)acrolein, 99%
3-(2-Furyl)acrolein, 99%
3-(2-Furyl)acrolein, 99%
Thermo Scientific Chemicals
3-(2-Furyl)acrolein, 99%
CAS: 623-30-3 | C7H6O2 | 122.123 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13042.1425 g
Catalog number ALFA13042.14
Price (MYR)
633.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
633.00
EA
Add to cart
Specifications
Chemical Name or Material3-(2-Furyl)acrolein
CAS623-30-3
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c