Nickel(II) acetate tetrahydrate, 98+%
Nickel(II) acetate tetrahydrate, 98+%
Nickel(II) acetate tetrahydrate, 98+%
Nickel(II) acetate tetrahydrate, 98+%
Thermo Scientific Chemicals
Nickel(II) acetate tetrahydrate, 98+%
CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA13026.36
Price (MYR)
527.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
527.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel(II) acetate tetrahydrate
CAS6018-89-9
Health Hazard 1H302+H332-H317-H334-H335-H341-H350i-H360D-H372
Health Hazard 2GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
View more
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nickel(II) acetate tetrahydrate is used in electroplating. It is also used in the preparation of polynuclear-nickel polyoxotungstate cluster compounds, which are used in designing molecular magnets. Further, it is used as a catalyst and mordant for dyeing textiles, as a sealant for anodized aluminum and to make other nickel compounds.
Solubility
Soluble in water and methanol. Insoluble in diethyl ether and n-octanol.
Notes
Hygroscopic. Incompatible with oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
  1. In combination with NaH and the Na derivative of 3-Hydroxy-1-methyl piperidine, L10829 , can be used for desufurization of thioethers, showing dramatic rate enhancement compared with other systems: Tetrahedron Lett., 39, 8987 (1998).
  2. Das, M.; Harms, K.; Ghosh, B. N.; Rissanen, K.; Chattopadhyay, S. Bis(μ-tetrazolato-NN') bridged dinuclear nickel(II) Schiff base complexes: Tandem synthesis, structure and self assembly. Polyhedron 2015, 87, 286-292.
  3. Sivakumar, P.; Nayak, P. K.; Markovsky, B.; Aurbach, D.; Gedanken, A. Sonochemical synthesis of LiNi0.5Mn1.5O4 and its electrochemical performance as a cathode material for 5V Li-ion batteries. Ultrason. Sonochem. 2015, 26, 332-339.