4-Methoxybenzhydrazide, 98+%
4-Methoxybenzhydrazide, 98+%
4-Methoxybenzhydrazide, 98+%
4-Methoxybenzhydrazide, 98+%
Thermo Scientific Chemicals
4-Methoxybenzhydrazide, 98+%
CAS: 3290-99-1 | C8H10N2O2 | 166.18 g/mol
Have Questions?
Catalog NumberQuantity
ALFA13015.0910 g
Catalog number ALFA13015.09
Price (MYR)
377.00
EA
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
377.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methoxybenzhydrazide
CAS3290-99-1
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point136°C to 140°C
View more
4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzhydrazide is used as pharmaceutical intermediates and it is also involved in a variety of organic synthesis.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Cheuk-Fai Chow; Shunsuke Fujii; Jean-Marie Lehn. Metallodynamers: neutral double-dynamic metallosupramolecular polymers. Chemistry An Asian Journal. 2008, 3, (8-9),1324-1335