trans-4-Nitrocinnamic acid, 98+%
trans-4-Nitrocinnamic acid, 98+%
trans-4-Nitrocinnamic acid, 98+%
trans-4-Nitrocinnamic acid, 98+%
Thermo Scientific Chemicals
trans-4-Nitrocinnamic acid, 98+%
CAS: 882-06-4 | C9H7NO4 | 193.158 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12990.1425 g
Catalog number ALFA12990.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-4-Nitrocinnamic acid
CAS882-06-4
Health Hazard 1H315-H319-H335-H341
Health Hazard 2GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
View more
trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
RUO – Research Use Only
General References:
  1. R Ketcham.; D Jambotkar. The preparation of cis-4-nitro-4'-methoxystilbene via the Wittig Reaction. J. Org. Chem. 196227, (12) , 4666-4667.
  2. X Yang.; SB Hall.; S Ngin Tan. Electrochemical reduction of a conjugated cinnamic acid diazonium salt as an immobilization matrix for glucose biosensor. Electroanalysis. 200315 (10) , 885-891.