1,2-Dimethoxyethane, 99+%, stab. with BHT
1,2-Dimethoxyethane, 99+%, stab. with BHT
1,2-Dimethoxyethane, 99+%, stab. with BHT
1,2-Dimethoxyethane, 99+%, stab. with BHT
1,2-Dimethoxyethane, 99+%, stab. with BHT
Thermo Scientific Chemicals
1,2-Dimethoxyethane, 99+%, stab. with BHT
CAS: 110-71-4 | C4H10O2 | 90.122 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
Catalog number ALFA12986.AP
Price (MYR)
472.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
472.00
EA
Add to cart
Specifications
Chemical Name or Material1,2-Dimethoxyethane
Name Notestabilized with BHT
CAS110-71-4
Health Hazard 1H225-H315-H319-H332-H360FD
Health Hazard 2GHS H Statement
H225-H360-H332
Highly flammable liquid and vapour.
May damage fertility or the unborn child.
Harmful if inhaled.
View more
1,2-Dimethoxyethane is widely used as a solvent for electrolyte of lithium batteries, polysilicones, oligo- and polysaccharides. It plays an important role in Grignard reactions, Suzuki reactions and Stille couplings in organometallic chemistry and in pharmaceutical synthesis. It is a higher boiling point solvent and is used as an alternative to diethyl ether and tetrahydrofuran. It is used for the etching of synthetic polymers like polytetrafluoroethylene and other fluoropolymers with alkali metal dispersions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Dimethoxyethane is widely used as a solvent for electrolyte of lithium batteries, polysilicones, oligo- and polysaccharides. It plays an important role in Grignard reactions, Suzuki reactions and Stille couplings in organometallic chemistry and in pharmaceutical synthesis. It is a higher boiling point solvent and is used as an alternative to diethyl ether and tetrahydrofuran. It is used for the etching of synthetic polymers like polytetrafluoroethylene and other fluoropolymers with alkali metal dispersions.
Solubility
Miscible with water, methanol, ethanol, diethyl ether, acetone, tetrahydrofuran, benzene and toluene.
Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It can be oxidized on air to form peroxides.
RUO – Research Use Only
General References:
  1. Solvent for lithiations and metal hydride reductions.
  2. Recommended solvent for the Schmidt reaction of ketones with hydrazoic acid, avoiding potential hazards reported with the more usual chlorinated solvents, particularly chloroform. The hydrazoic acid is generated in situ from NaN3 and methanesulfonic acid: Tetrahedron, 52, 1609 (1996).
  3. Che, P.; Lu, F.; Si, X.; Xu, J. Catalytic etherification of hydroxyl compounds to methyl ethers with 1,2-dimethoxyethane. RSC Adv. 2015, 5 (31), 24139-24143.
  4. Monreal, M. J.; Thomson, R. K.; Scott, B. L.; Kiplinger, J. L. Enhancing the synthetic efficacy of thorium tetrachloride bis(1,2-dimethoxyethane) with added 1,2-dimethoxyethane: Preparation of metallocene thorium dichlorides. Inorg. Chem. Commun. 2014, 46, 51-53.