4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
Thermo Scientific Chemicals
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
CAS: 2991-42-6 | C7H4ClF3O2S | 244.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA12907.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Trifluoromethyl)benzenesulfonyl chloride
CAS2991-42-6
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c