Potassium hydrogen phthalate, 99%
Potassium hydrogen phthalate, 99%
Potassium hydrogen phthalate, 99%
Potassium hydrogen phthalate, 99%
Thermo Scientific Chemicals
Potassium hydrogen phthalate, 99%
CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
1000 g
Catalog number ALFA12895.0I
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hydrogen phthalate
CAS877-24-7
Melting Point297°C (decomposition)
Recommended StorageAmbient temperatures
Density1.636
View more
Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters and for the preparation of volumetric alkali solutions. It acts as buffer in pH determination. It finds application as a useful standard for total organic carbon testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters and for the preparation of volumetric alkali solutions. It acts as buffer in pH determination. It finds application as a useful standard for total organic carbon testing.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kahr, B.; Bullard, T.; Kurimoto, M.; Yoreo, J. J. Complex organic guests in simple crystals: potassium hydrogen phthalate as case study. Cryst. Res. Technol. 2013, 48 (10), 849-863.
  2. Sudhahar, S.; Kumar, M. K.; Jayaramakrishnan, V.; Muralidharan, R.; Kumar, R. M. Effect of Sm+ Rare Earth Ion on the Structural, Thermal, Mechanical and Optical Properties of Potassium Hydrogen Phthalate Single Crystals. J. Mater. Sci. Technol. 2014, 30 (1), 13-18.