2-Methylcyclohexanol, cis + trans, 97%
2-Methylcyclohexanol, cis + trans, 97%
2-Methylcyclohexanol, cis + trans, 97%
2-Methylcyclohexanol, cis + trans, 97%
Thermo Scientific Chemicals
2-Methylcyclohexanol, cis + trans, 97%
CAS: 583-59-5 | C7H14O | 114.188 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12877.AP500 mL
Catalog number ALFA12877.AP
Price (MYR)
696.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
696.00
EA
Add to cart
Specifications
Chemical Name or Material2-Methylcyclohexanol
Name Notecis + trans
CAS583-59-5
Health Hazard 1H226-H302+H332-H319
Health Hazard 2GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.
View more
2-Methylcyclohexanol is used in the preparation of acetic acid-(2-methyl-cyclohexyl ester) by reaction with acetic anhydride. Further, it is used to study the effect of organic solvents on epoxide hydrolase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methylcyclohexanol is used in the preparation of acetic acid-(2-methyl-cyclohexyl ester) by reaction with acetic anhydride. Further, it is used to study the effect of organic solvents on epoxide hydrolase.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Shafaghat, H.; Rezaei, P. S.; Daud, W. M. A. W. Catalytic hydrogenation of phenol, cresol and guaiacol over physically mixed catalysts of Pd/C and zeolite solid acids. RSC Adv. 2015, 5 (43), 33990-33998.
  2. Takai, Y.; Touhara, K. Enantioselective recognition of menthol by mouse odorant receptors. Biosci. Biotechnol. Biochem. 2015, 79 (12), 1980-1986.