2,4-Dihydroxybenzaldehyde, 98%
2,4-Dihydroxybenzaldehyde, 98%
2,4-Dihydroxybenzaldehyde, 98%
2,4-Dihydroxybenzaldehyde, 98%
Thermo Scientific Chemicals
2,4-Dihydroxybenzaldehyde, 98%
CAS: 95-01-2 | C7H6O3 | 138.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFA12868.22
Price (MYR)
1,332.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,332.00
EA
Add to cart
Specifications
Chemical Name or Material2,4-Dihydroxybenzaldehyde
CAS95-01-2
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dihydroxybenzaldehyde is used in two-step synthesis of ethyl 3,5-dibromo-2,4-dihydroxycinnamate. It has a potent antioxidative and antibacterial activity.
Solubility
Soluble in water. Solubility in methanol is almost transparent. Easily soluble in alcohol, ethyl ether, chloroform, glacial acetic acid. Insoluble in benzene and easily decomposes under strong acid strong alkali.
Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
  1. Nathalie Gagey, et al. Two-photon uncaging with the efficient 3,5-dibromo-2,4-dihydroxycinnamic caging group.Angew Chem Int Ed Engl.,2007,46(14), 2467-2469.
  2. Mendelson WL, et al. The regioselective 4-benzylation of 2, 4-dihydroxybenzaldehyde.Synth. Commun.,1996,26(3), 593-601.