Behenic acid, tech. 85%
Behenic acid, tech. 85%
Behenic acid, tech. 85%
Behenic acid, tech. 85%
Thermo Scientific Chemicals
Behenic acid, tech. 85%
CAS: 112-85-6 | C22H44O2 | 340.592 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12850.30250 g
Catalog number ALFA12850.30
Price (MYR)
273.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
273.00
EA
Add to cart
Specifications
Chemical Name or MaterialBehenic acid
CAS112-85-6
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.
Solubility
Soluble in DMF (∼3 mg/ml), hot methanol, water (0.15 mg/ml at 25°C), chloroform, and ethanol (2.18 mg/ml at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, reducing agents and bases. Store at room temperature.
RUO – Research Use Only
General References:
  1. M. Paudler.; J. Ruths.; H. Riegler. Analysis of multiple-angle ellipsometry of uniaxial ultrathin organic films at the air-water interface and determination of the refractive indices of behenic acid monolayers. Langmuir. 1992, 8 (1), 184-189.
  2. Bing Guan.; Zhimin Liu.; Buxing Han.; Haike Yan. Solubility of behenic acid in supercritical carbon dioxide with ethanol. The Journal of Supercritical Fluids. 1999, 14 (3), 213-218.