Potassium sorbate, 99%
Potassium sorbate, 99%
Potassium sorbate, 99%
Potassium sorbate, 99%
Thermo Scientific Chemicals
Potassium sorbate, 99%
CAS: 24634-61-5 | C6H7KO2 | 150.218 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number ALFA12844.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium Sorbate
CAS24634-61-5
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Potassium sorbate serves as an inhibitor for molds, yeasts in many food items like cheese, wine, yogurt, dried meats, apple cider, soft drinks, fruit drinks and baked goods. It is used in the production of syrup, milkshakes, creams, lotions, shampoos, makeup and sunscreen products. It is mainly used to increase the shelf life of various commercial products without affecting its taste, smell and color of the food. It is used as a preservative and considered to be a replacement for parabens.
Solubility
Soluble in water, alcohol, propylene glycol. Slightly soluble in acetoneand chloroform, corn oil and ether. Insoluble in benzene.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Hafner, S. D.; Windle, M.; Merrill, C.; Smith, M. L.; Franco, R. B.; Kung, L. Effects of potassium sorbate and Lactobacillus plantarum MTD1 on production of ethanol and other volatile organic compounds in corn silage. Anim. Feed Sci. Technol. 2015, 208, 79-85.
  2. Amirpour, M.; Arman, A.; Yolmeh, A.; Azam, M. A.; Sodium benzoate and potassium sorbate preservatives in food stuffs in Iran. Food Addit. Contam. B 2015, 8 (2), 142-148.