trans-4-Methoxycinnamaldehyde, 98+%
trans-4-Methoxycinnamaldehyde, 98+%
trans-4-Methoxycinnamaldehyde, 98+%
trans-4-Methoxycinnamaldehyde, 98+%
trans-4-Methoxycinnamaldehyde, 98+%
CAS: 24680-50-0 | C10H10O2 | 162.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12839.0910 g
Catalog number ALFA12839.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-4-Methoxycinnamaldehyde
CAS24680-50-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-4-Methoxycinnamaldehyde is used as an intermediate in chemical research.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Dr. Marco Radi.; Dr. Mafalda Pagano.; Dr. Luigi Franchi.; Dr. Daniele Castagnolo.; Prof. Silvia Schenone.; Dr. Gianni Casaluce.; Dr. Claudio Zamperini.; Dr. Elena Dreassi.; Dr. Giovanni Maga.; Dr. Alberta Samuele.; Dr. Encarna Gonzalo.; Dr. Bonaventura Clotet.; Prof. José A. Esté.;Prof. Maurizio Botta. Synthesis, Biological Activity, and ADME Properties of Novel S-DABOs-DABOs as HIV Reverse Transcriptase Inhibitors. ChemMedChem. 2012, 7 (5), 883-896.
  2. A. Asadi Sardari.; A.A. Hojat Jalali.; S. Bahraminejad.; D. Safaee. Effect of plant extracts on the mortality of root-knot nematodes’ J2, Meloidogyne javanica. Archives of Phytopathology and Plant Protection. 2015, 15 (4), 365-375.