Tetramethylammonium iodide, 99%
Tetramethylammonium iodide, 99%
Tetramethylammonium iodide, 99%
Tetramethylammonium iodide, 99%
Thermo Scientific Chemicals
Tetramethylammonium iodide, 99%
CAS: 75-58-1 | C4H12IN | 201.05 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12811.22100 g
Catalog number ALFA12811.22
Price (MYR)
1,090.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
1,090.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetramethylammonium iodide
CAS75-58-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c