Bromoethane, 98%
Bromoethane, 98%
Bromoethane, 98%
Thermo Scientific Chemicals
Bromoethane, 98%
CAS: 74-96-4 | C2H5Br | 108.966 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA12804.36
Price (MYR)
355.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
355.00
EA
Add to cart
Specifications
Chemical Name or MaterialBromoethane
CAS74-96-4
Health Hazard 1H225-H302+H332-H351
Health Hazard 2GHS H Statement
H225-H351-H302-H332
Health Hazard 3P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P330-P370+P378q-P501c
View more
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.
Solubility
Miscible with ethanol, ether, chloroform and organic solvents.
Notes
Light sensitive. Store in cool place. Incompatible with strong bases, strong oxidizing agents, magnesium, potassium, calcium, sodium oxides, aluminum, zinc and alkali metals.
RUO – Research Use Only
General References:
  1. Sarzyński, D.; Gola, A. A.; Dryś, A.; Brudnik, K.; Jodkowski, J. T. Kinetic study of the reaction of chlorine atoms with bromoethane and D-bromoethane in the gas phase. Chem. Phys. Lett. 2011, 509 (4-6), 114-118.
  2. Zhu, J.; Zhang, Y.; Tian, M.; Liu, J. Fabrication of a Mixed Matrix Membrane with in Situ Synthesized Quaternized Polyethylenimine Nanoparticles for Dye Purification and Reuse. ACS Sustainable Chem. Eng. 2015, 3 (4), 690-701.