3-(Trifluoromethyl)phenol, 98+%
3-(Trifluoromethyl)phenol, 98+%
3-(Trifluoromethyl)phenol, 98+%
3-(Trifluoromethyl)phenol, 98+%
Thermo Scientific Chemicals
3-(Trifluoromethyl)phenol, 98+%
CAS: 98-17-9 | C7H5F3O | 162.111 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12784.1425 g
Catalog number ALFA12784.14
Price (MYR)
350.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
350.00
EA
Add to cart
Specifications
Chemical Name or Material3-(Trifluoromethyl)phenol
CAS98-17-9
Health Hazard 1H227-H302-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H302-H312-H332-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Health Hazard 3P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c