Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Ethyl 4-aminobenzoate, 98%
Thermo Scientific Chemicals
Ethyl 4-aminobenzoate, 98%
CAS: 94-09-7 | C9H11NO2 | 165.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA12754.30
Price (MYR)
565.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
565.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl 4-aminobenzoate
CAS94-09-7
Health Hazard 1H315-H317-H319
Health Hazard 2GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
View more
Ethyl 4-aminobenzoate is used as an anesthetic and utilized for topical pain reliever. It is mixed with antipyrine to get A/B otic drops, which is used in the removal of earwax as well as relieve ear from pain. It exhibits analgesic and antipruritic effect mainly used for wounds, ulcers, mucosal surfaces and hemorrhoids narcotic pain. Further, it is a main constituent of anesthetic ointments. In addition to this, it is effective to toothache, sore throat, oral ulcers, hemorrhoids and anal fissure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 4-aminobenzoate is used as an anesthetic and utilized for topical pain reliever. It is mixed with antipyrine to get A/B otic drops, which is used in the removal of earwax as well as relieve ear from pain. It exhibits analgesic and antipruritic effect mainly used for wounds, ulcers, mucosal surfaces and hemorrhoids narcotic pain. Further, it is a main constituent of anesthetic ointments. In addition to this, it is effective to toothache, sore throat, oral ulcers, hemorrhoids and anal fissure.
Solubility
Soluble in ethanol, chloroform, ethyl ether and dilute acids. Sparingly soluble in water
Notes
Incompatible with strong oxidizing agents, reducing agents, acids and bases.
RUO – Research Use Only
General References:
  1. Warnke, S.; Seo, J.; Boschmans, J.; Sobott, F.; Scrivens, J. H.; Bleiholder, C.; Bowers, M. T.; Gewinner, S.; Schöllkopf, W.; Pagel, K.; von Helden, G. Protomers of Benzocaine: Solvent and Permittivity Dependence. J. Am. Chem. Soc. 2015, 137 (12), 4236-4242.
  2. Alotaibi, M. R.; Husbands, S. M.; Blagbrough, I. S. 1H, 13C, 15N HMBC, and 19F NMR spectroscopic characterisation of seized flephedrone, cut with benzocaine. J. Pharm. Biomed. Anal. 2015, 107, 535-538.