1-Dodecanethiol, 98%
1-Dodecanethiol, 98%
1-Dodecanethiol, 98%
1-Dodecanethiol, 98%
Thermo Scientific Chemicals
1-Dodecanethiol, 98%
CAS: 112-55-0 | C12H26S | 202.40 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12741.22100 g
Catalog number ALFA12741.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Dodecanethiol
CAS112-55-0
Health Hazard 1H314-H317-H335
Health Hazard 2GHS H Statement
H315-H319-H317-H335
Health Hazard 3P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
View more
1-Dodecanethiol is used in the preparation of hydrophobic or mixed self-assembled monolayers. It is also employed as a chain transfer agent for radical polymerization. Further, it is utilized as a polymerization inhibitor in polyurethane and neoprene adhesives, which finds application in the footwear industry. In addition to this, it acts as a protein regenerating agent used for the regeneration of native proteins from mercuribenzoate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Dodecanethiol is used in the preparation of hydrophobic or mixed self-assembled monolayers. It is also employed as a chain transfer agent for radical polymerization. Further, it is utilized as a polymerization inhibitor in polyurethane and neoprene adhesives, which finds application in the footwear industry. In addition to this, it acts as a protein regenerating agent used for the regeneration of native proteins from mercuribenzoate.
Solubility
Immiscible with water.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
RUO – Research Use Only
General References:
  1. Chen, D.; Cui, P.; Cao, H.; Yang, J. A 1-dodecanethiol-based phase transfer protocol for the highly efficient extraction of noble metal ions from aqueous phase. J. Env.Sci. 2015, 29, 146-150.
  2. Dickson, R. E.; Hu, M. Z. Chemical synthesis and optical characterization of regular and magic-sized CdS quantum dot nanocrystals using 1-dodecanethiol. J. Mater. Res. 2015, 30 (7), 890-895.