Polyvinyl acetate, M.W. ca 50,000, Thermo Scientific™
Polyvinyl acetate, M.W. ca 50,000, Thermo Scientific™
Polyvinyl acetate, M.W. ca 50,000, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA12732.36
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyvinyl acetate
CAS9003-20-7
Recommended StorageAmbient temperatures
TSCAYes
Molecular Formula(C4H6O2)n
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.