4-Hydroxybenzhydrazide, 98%
4-Hydroxybenzhydrazide, 98%
4-Hydroxybenzhydrazide, 98%
4-Hydroxybenzhydrazide, 98%
Thermo Scientific Chemicals
4-Hydroxybenzhydrazide, 98%
CAS: 5351-23-5 | C7H8N2O2 | 152.153 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12702.1425 g
Catalog number ALFA12702.14
Price (MYR)
344.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
344.00
EA
Add to cart
Specifications
Chemical Name or Material4-Hydroxybenzhydrazide
CAS5351-23-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c