Tetra-n-butylammonium hydroxide, 40% w/w in methanol
Tetra-n-butylammonium hydroxide, 40% w/w in methanol
Tetra-n-butylammonium hydroxide, 40% w/w in methanol
Tetra-n-butylammonium hydroxide, 40% w/w in methanol
Thermo Scientific Chemicals
Tetra-n-butylammonium hydroxide, 40% w/w in methanol
CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2500 mL
Catalog number ALFA12626.AP
Price (MYR)
1,631.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,631.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetra-n-butylammonium hydroxide
CAS2052-49-5
Health Hazard 1H225-H301+H311+H331-H314-H335-H370
Health Hazard 2GHS H Statement
H225-H301-H311-H331-H370-H314-H318
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more
Tetra-n-butylammonium hydroxide, 40% w/w in methanol is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is also used as a curing accelerator for epoxy resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-butylammonium hydroxide, 40% w/w in methanol is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is also used as a curing accelerator for epoxy resins.
Solubility
Miscible with organic solvents.
Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
RUO – Research Use Only
General References:
  1. Bauer, M.; Spange, S. Enhancing the Reactivity of an Electrophilic Barbiturate Dye by Cooperative Hydrogen Bonding. Eur. J. Org. Chem. 2010, 2010 (2), 259-264.
  2. Clark, D. C.; Baker, W. E.; Russell, K. E.; Whitney, R. A. Dual monomer grafting of styrene and maleic anhydride onto model hydrocarbon substrates. J. Polym. Sci. A Polym. Chem. 2000, 38 (13), 2456-2468.