(+/-)-Camphor-10-sulfonic acid, 98%
(+/-)-Camphor-10-sulfonic acid, 98%
(+/-)-Camphor-10-sulfonic acid, 98%
(+/-)-Camphor-10-sulfonic acid, 98%
Thermo Scientific Chemicals
(+/-)-Camphor-10-sulfonic acid, 98%
CAS: 5872-08-2 | C10H16O4S | 232.29 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12620.22100 g
Catalog number ALFA12620.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-Camphor-10-sulfonic acid
CAS5872-08-2
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
(+)-Camphor-10-sulfonic acid is used as a resolving agent for chiral amines and other cations. It is involved in the preparation of active pharmaceutical ingredient such as trimetaphan camsilate, which is used to reduce bleeding during neurosurgery. Further, it is involved in the transglucosidation of methyl and ethyl D-glucopyranosides by alcoholysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+)-Camphor-10-sulfonic acid is used as a resolving agent for chiral amines and other cations. It is involved in the preparation of active pharmaceutical ingredient such as trimetaphan camsilate, which is used to reduce bleeding during neurosurgery. Further, it is involved in the transglucosidation of methyl and ethyl D-glucopyranosides by alcoholysis.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Useful acid catalyst, giving high yields in the formation of THP ethers from alcohols; see, e.g.: J. Chem. Soc., Chem. Commun., 413 (1986); Org. Synth., 76, 178 (1998). Catalyst for cycloprotection of 1,2-diols with 1,2-Cyclohexanedione, A14401: Synlett, 793 (1996); Org. Synth., 75, 170 (1997). Preferred catalyst for the intramolecular cyclization of hydroxy epoxides to tetrahydrofurans or tetrahydropyrans: J. Am. Chem. Soc., 111, 5330 (1989).
  2. Chandam, D. R.; Mulik, A. G.; Patil, P. P.; Jagdale, S. D.; Patil, D. R.; Deshmukh, M. B. (±)-Camphor-10-sulfonic acid catalyzed atom efficient and green synthesis of triazolo[1,2-a]indazole-triones and spiro triazolo[1,2-a]indazole-tetraones. Res. Chem. Intermed. 2015, 41 (2), 761-771.
  3. Shelke, A. M.; Suryavanshi, G. An efficient organocatalytic route for asymmetric total synthesis of Stagonolide F. Tetrahedron Lett. 2015, 56 (45), 6207-6209.