2-Fluoroaniline, 99%
2-Fluoroaniline, 99%
2-Fluoroaniline, 99%
2-Fluoroaniline, 99%
Thermo Scientific Chemicals
2-Fluoroaniline, 99%
CAS: 348-54-9 | C6H6FN | 111.12 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12612.22100 g
Catalog number ALFA12612.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Fluoroaniline
CAS348-54-9
Health Hazard 1H226-H302+H312+H332-H315-H318-H335
Health Hazard 2GHS H Statement
H331-H226-H373-H302-H312-H315-H319
Toxic if inhaled.
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c