Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals
Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals
Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals
Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals
Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFA12593.18
Price (MYR)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-n-hexyl phthalate
CAS84-75-3
Health Hazard 1H360FD
Health Hazard 2GHS H Statement
H360
May damage fertility or the unborn child.
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phthalates are by far the most widely used plasticizers, primarily to make soft and flexible polyvinyl chloride (PVC) for the applications in the industry of automotive, building & construction material, cable, flooring, medical device and toys. Minor quantity of phthalates are used in adhesives, caulk, sealants, paint to improve work performance. Small molecule phthalates are used as solvents in perfumes to provide longer linger longer and in nail polish to prevent chipping.
Solubility
Soluble in water (0.24 mg/L).
Notes
Store at 4°C. Avoid excessive heat and light. Reactive towards oxidizing agents.
RUO – Research Use Only
General References:
  1. Alan H. Mann.; Shirley C. Price.; Fiona E. Mitchell.; Paul Grasso.; Richard H. Hinton.; James W. Bridges. Comparison of the short-term effects of di(2-ethylhexyl) phthalate, di(n-hexyl) phthalate, and di(n-octyl) phthalate in rats. Toxicol. Appl. Pharmacol. 1985, 77 (1),116-132.
  2. Paul M.D. Foster.; Lucy V. Thomas.; Melvyn W. Cook.; Sharat D. Gangolli. Study of the testicular effects and changes in zinc excretion produced by some n-alkyl phthalates in the rat. Toxicol. Appl. Pharmacol. 1980, 54 (3), 392-398.