Phenoxyacetic acid, 98%
Phenoxyacetic acid, 98%
Phenoxyacetic acid, 98%
Phenoxyacetic acid, 98%
Thermo Scientific Chemicals
Phenoxyacetic acid, 98%
CAS: 122-59-8 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA12541.36
Price (MYR)
495.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
495.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenoxyacetic acid
CAS122-59-8
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
Phenoxyacetic acid is a flavoring ingredient. Its photodegradation using titanium dioxide as photocatalyst has been studied. Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenoxyacetic acid is a flavoring ingredient. Its photodegradation using titanium dioxide as photocatalyst has been studied. Selective separation of penicillin V from phenoxyacetic acid using liquid membranes consisting of 1,2-dichloroethane and Amberlite LA-2 as carrier has been studied.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Robert M.Muir; Corwin Hansch. The relationship of structure and plant-growth activity of substituted benzoic and phenoxyacetic acids. Plant Physiol. 1951, 26, (2)369-374
  2. A.R.Harker; R.H.Olsen; R.J.Seidler. Phenoxyacetic acid degradation by the 2,4-dichlorophenoxyacetic acid (TFD) pathway of plasmid pJP4: mapping and characterization of the TFD regulatory gene, tfdR. J. Bacteriol. 1989, 171, (1)314-320