3-Acetylcoumarin, 98+%
3-Acetylcoumarin, 98+%
3-Acetylcoumarin, 98+%
3-Acetylcoumarin, 98+%
Thermo Scientific Chemicals
3-Acetylcoumarin, 98+%
CAS: 3949-36-8 | C11H8O3 | 188.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA12532.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Acetylcoumarin
CAS3949-36-8
Melting Point119°C to 124°C
Recommended StorageAmbient temperatures
EINECS Number223-541-6