Methyl dodecanoate, 99%
Methyl dodecanoate, 99%
Methyl dodecanoate, 99%
Methyl dodecanoate, 99%
Thermo Scientific Chemicals
Methyl dodecanoate, 99%
CAS: 111-82-0 | C13H26O2 | 214.349 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALFA12492.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl dodecanoate
CAS111-82-0
Melting Point4°C to 5°C
Recommended StorageAmbient temperatures
DOT InformationHazard Class: 9; Packaging Group: III
View more
Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl dodecanoate used in production of biodiesel. It has been used in selective synthesis of the secondary amide surfactant, N-methyl lauroylethanolamide. It is also utilized as emollients, perfuming agents skin conditioning.
Solubility
Not miscible in water. Soluble in alcohol, dipropylene glycol.
Notes
Store at room temperature. Ensure good ventilation.
RUO – Research Use Only
General References:
  1. Beata Korzystka.; Hanna Adamczak.; Anna Sobczyńska.; Jan Szymanowski. Ultrafiltration characteristics of colloid solutions containing oxyethylated methyl dodecanoate, hexadecyltrimethylammonium bromide and selected phenols as pollutants. Colloids Surf., A. 2003, 212 (2-3),175-183.
  2. I. Hama.; T. Okamoto.; E. Hidai.; K. Yamada. Direct ethoxylation of fatty methyl ester over Al-Mg composite oxide catalyst. J. Am. Oil Chem. Soc. 1997, v (1),19-24.