5-Hydroxymethyl-2-furaldehyde, 97%
5-Hydroxymethyl-2-furaldehyde, 97%
5-Hydroxymethyl-2-furaldehyde, 97%
5-Hydroxymethyl-2-furaldehyde, 97%
Thermo Scientific Chemicals
5-Hydroxymethyl-2-furaldehyde, 97%
CAS: 67-47-0 | C6H6O3 | 126.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA12475.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Hydroxymethyl-2-furaldehyde
CAS67-47-0
Health Hazard 1H227-H315-H319-H335-H373
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P235-P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P370+P378q-P501c
View more
5-Hydroxymethyl-2-furaldehyde is used as an intermediate of OMBF (5,5'-oxydimethylenebis(2- furfural)), crown ethers. It is used in organic synthesis to prepare dialdehydes, glycols, ethers, amino alcohols and acetals. It acts as an antioxidant found in some fruit juices and caramel products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Hydroxymethyl-2-furaldehyde is used as an intermediate of OMBF (5,5′-oxydimethylenebis(2- furfural)), crown ethers. It is used in organic synthesis to prepare dialdehydes, glycols, ethers, amino alcohols and acetals. It acts as an antioxidant found in some fruit juices and caramel products.
Solubility
Soluble in water, alcohol, ethyl acetate, acetone, dimethylformamide, benzene, ether and chloroform.
Notes
Air and light sensitive. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
  1. Demirhan, B. E.; Demirhan, B.; Sonmez, C.; Torul, H.; Tamer, U.; Yentur, G. Short communication: Determination of potential 5-hydroxymethyl-2-furaldehyde and 2-furaldehyde compounds in follow-on milks and infant formulas using the high-performance liquid chromatography method. J. Dairy Sci. 2015, 98 (2), 818-822.
  2. Lee, Y.; Gao, Q.; Kim, E.; Lee, Y.; Park, S. J.; Lee, H. E.; Jang, D. S.; Ryu, J. H. Pretreatment with 5-hydroxymethyl-2-furaldehyde blocks scopolamine-induced learning deficit in contextual and spatial memory in male mice. Pharmacol. Biochem. Behav 2015, 134, 57-64.