Ammonium iron(II) sulfate hexahydrate, 99%
Ammonium iron(II) sulfate hexahydrate, 99%
Ammonium iron(II) sulfate hexahydrate, 99%
Ammonium iron(II) sulfate hexahydrate, 99%
Ammonium iron(II) sulfate hexahydrate, 99%
Thermo Scientific Chemicals
Ammonium iron(II) sulfate hexahydrate, 99%
CAS: 7783-85-9 | FeH20N2O14S2 | 392.13 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12473.36500 g
Catalog number ALFA12473.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium iron(II) sulfate hexahydrate
Melting Point∼100°C (decomposition)
CAS7783-85-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Ammonium iron(II) sulfate hexahydrate is widely utilized in analytical chemistry for titration purposes since it is affected lesser by oxygen in the air than iron(II) sulpahte. It is employed to measure high doses of gamma rays by using Fricke's dosemeter. It is involved in a variety of other applications from nanomaterials to general redox reactions. It is used in the medical field as an antianemic agent for treatment of iron deficiency.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Air and light sensitive. Incompatible with strong acids and oxidizing agents.
RUO – Research Use Only
General References:
  1. Uchida, M.; Qazi, S.; Edwards, E.; Douglas, T. Use of Protein Cages as a Template for Confined Synthesis of Inorganic and Organic Nanoparticles. Methods Mol. Biol. 2014, 1252, 17-25.
  2. Karvinen, A.; Lehtinen, L.; Kankaala, P. Variable Effects of Iron (Fe (III)) Additions on Potential Methane Production in Boreal Lake Littoral Sediments. Wetlands 2015, 35 (1), 137-146.