2,4-Dichlorophenoxyacetic acid, 98%
2,4-Dichlorophenoxyacetic acid, 98%
2,4-Dichlorophenoxyacetic acid, 98%
2,4-Dichlorophenoxyacetic acid, 98%
Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 98%
CAS: 94-75-7 | C8H6Cl2O3 | 221.033 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALFA12467.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material2,4-Dichlorophenoxyacetic acid
CAS94-75-7
Health Hazard 1H302-H317-H318-H335-H351
Health Hazard 2GHS H Statement
H318-H302-H312-H317-H335
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P501c
View more
2,4-Dichlorophenoxyacetic acid is used as a supplement in plant cell culture media. It is a synthetic auxin, which finds application as a plant growth regulator. Further, it is used to increase latex output of old rubber trees.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dichlorophenoxyacetic acid is used as a supplement in plant cell culture media. It is a synthetic auxin, which finds application as a plant growth regulator. Further, it is used to increase latex output of old rubber trees.
Solubility
Slightly soluble in water.
Notes
Moisture and light-sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Loomis, D.; Guyton, K.; Grosse, Y.; El Ghissasi, F.; Bouvard, V.; Benbrahim-Tallaa, L.; Guha, N.; Mattock, H.; Straif, K. Carcinogenicity of lindane, DDT, and 2,4-dichlorophenoxyacetic acid. Lancet Oncol. 2015, 16 (8), 891-892.
  2. Njoku, V. O.; Islam, M. A.; Asif, M.; Hameed, B. H. Adsorption of 2,4-dichlorophenoxyacetic acid by mesoporous activated carbon prepared from H3PO4-activated langsat empty fruit bunch. J. Environ. Manage. 2015, 154, 138-144.