2-Aminopyridine, 99%
2-Aminopyridine, 99%
2-Aminopyridine, 99%
2-Aminopyridine, 99%
Thermo Scientific Chemicals
2-Aminopyridine, 99%
CAS: 504-29-0 | C5H6N2 | 94.117 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Catalog number ALFA12374.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Aminopyridine
CAS504-29-0
Health Hazard 1H301-H312-H315-H319-H500
Health Hazard 2GHS H Statement
H301-H331-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.
Solubility
Slightly soluble in water(8g/100 mL at 19°C)
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
RUO – Research Use Only
General References:
  1. M. Chao,; E. Schemp,; R. D. Rosenstein. 2-Aminopyridine. Acta Cryst. 1975, B31(12),2922-2924
  2. Zhou Jie,; He Xiwen.Study of the nature of recognition in molecularly imprinted polymer selective for 2-aminopyridine. Analytica Chimica Acta. 1999, 381(1),85-91.