Sulfadiazine, 98%
Sulfadiazine, 98%
Sulfadiazine, 98%
Thermo Scientific Chemicals
Sulfadiazine, 98%
CAS: 68-35-9 | C10H10N4O2S | 250.276 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12370.1850 g
Catalog number ALFA12370.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfadiazine
CAS68-35-9
Health Hazard 1H302-H315-H317-H319-H334-H335
Health Hazard 2GHS H Statement
H334-H302-H335-H315-H319-H317
Health Hazard 3P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
View more
Sulfadiazine is used as an antibacterial. It is a short-acting sulfonamide that is commonly used with pyrimethamine to treat toxoplasmosis in patients with acquired immunodeficiency syndrome. It is also used to treat newborns with congenital infections and to control acute infections when studying murine models of reactivated toxoplasmosis. It is It eliminates bacteria that cause infections by stopping the production of folic acid inside the bacterial cell, and is commonly used to treat urinary tract infections (UTIs)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Charles L. Fox Jr.; Shanta M. Modak. Mechanism of Silver Sulfadiazine Action on Burn Wound Infections. Antimicrob. Agents Chemother. 1974, 5 (6), 582-588.
  2. Shanta M. Modak.; Charles L. Fox Jr. Binding of silver sulfadiazine to the cellular components of Pseudomonas aeruginosa. Biochemical Pharmacology. 1973, 22 (19), 2391-2404.