1,10-Dibromodecane, 97%
1,10-Dibromodecane, 97%
1,10-Dibromodecane, 97%
1,10-Dibromodecane, 97%
Thermo Scientific Chemicals
1,10-Dibromodecane, 97%
CAS: 4101-68-2 | C10H20Br2 | 300.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12357.22100 g
Catalog number ALFA12357.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1,10-Dibromodecane
CAS4101-68-2
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sea
RUO – Research Use Only
General References:
  1. V. Percec.; M. Kawasumi.; P. L. Rinaldi.; V. E. Litman. Synthesis and characterization of cyclic liquid crystalline oligomers based on 1-(4-hydroxy-4'-biphenylyl)-2-(4-hydroxyphenyl)butane and 1,10-dibromodecane. Macromolecules. 1992, 25 (15), 3851-3861.
  2. Abil E. Aliev.; Sharon P. Smart.; Ian J. Shannon.; Kenneth D. M. Harris. Structural and dynamic properties of the 1,10-dibromodecane/urea inclusion compound, investigated by variable-temperature powder X-ray diffraction, solid-state 2H NMR lineshape analysis and solid-state 2H NMR spin-lattice relaxation time measurements. J. Chem. Soc., Faraday Trans. 1992, 96 (12), 2179-2185.