4-Nitrophenyl phosphate disodium salt hexahydrate, 96%
4-Nitrophenyl phosphate disodium salt hexahydrate, 96%
4-Nitrophenyl phosphate disodium salt hexahydrate, 96%
4-Nitrophenyl phosphate disodium salt hexahydrate, 96%
Thermo Scientific Chemicals
4-Nitrophenyl phosphate disodium salt hexahydrate, 96%
CAS: 333338-18-4 | C6H4NNa2O6P | 263.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA12310.14
Price (MYR)
1,193.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,193.00
EA
Add to cart
Specifications
Chemical Name or Material4-Nitrophenyl phosphate disodium salt hexahydrate
CAS333338-18-4
Melting Point>300°C
Recommended StorageKeep cold
EINECS Number224-246-5
View more
Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.
Solubility
Miscible with water. Immiscible with ethanol and acetone.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
  1. Chromogenic substrate for phosphatases: Methods of Enzymatic Analysis, 3rd ed., H. U. Bergmeyer, Ed., Verlag Chemie, Weinheim (1984), vol. 4, p76.
  2. Lee, J.; Yoo, J. K.; Sohn, H. J.; Kang, H. K.; Kim, D.; Shin, H. J.; Kim, J. H. Protective immunity against Naegleria fowleri infection on mice immunized with the rNfa1 protein using mucosal adjuvants. Parasitol. Res. 2015, 114 (4), 1377-1385.
  3. Du, D.; Ushida, T.; Furukawa, K. S. Influence of cassette design on three-dimensional perfusion culture of artificial bone. J. Biomed. Mater. Res. Part B Appl. Biomater. 2015, 103 (1), 84-91.