1-Bromo-2,4-dichlorobenzene, 98%
1-Bromo-2,4-dichlorobenzene, 98%
1-Bromo-2,4-dichlorobenzene, 98%
1-Bromo-2,4-dichlorobenzene, 98%
Thermo Scientific Chemicals
1-Bromo-2,4-dichlorobenzene, 98%
CAS: 1193-72-2 | C6H3BrCl2 | 225.894 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12298.065 g
Catalog number ALFA12298.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromo-2,4-dichlorobenzene
CAS1193-72-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4'-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-2,4-dichlorobenzene is used to produce 2,4,4′-trichloro-biphenyl. This reaction will need aq. Na2CO3 and solvents dioxane, ethanol. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Jeanne L. Bollige.; Christian M. Frech. The 1,3-Diaminobenzene-Derived Aminophosphine Palladium Pincer Complex {C6H3[NHP(piperidinyl)2]2Pd(Cl)} - A Highly Active Suzuki-Miyaura Catalyst with Excellent Functional Group Tolerance. Advanced Synthesis & Catalysis. 2010, 352 (6), 1075-1080.
  2. Okada, Y.; Yokozawa, M.;Akiba, M., Oishi, K.; Kyoji, O.; Akeboshi, T.; Nishimura, J. Bromination by means of sodium monobromoisocyanurate (SMBI). Organic & biomolecular chemistry. 2003, 1 (14), 2506-2511.