Benzhydrazide, 98%
Benzhydrazide, 98%
Benzhydrazide, 98%
Thermo Scientific Chemicals
Benzhydrazide, 98%
CAS: 613-94-5 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA12268.36
Price (MYR)
1,458.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,458.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzhydrazide
CAS613-94-5
Health Hazard 1H302-H315-H319-H335-H351
Health Hazard 2GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c