4-(4-Nitrophenylazo)resorcinol, 90+%
4-(4-Nitrophenylazo)resorcinol, 90+%
4-(4-Nitrophenylazo)resorcinol, 90+%
4-(4-Nitrophenylazo)resorcinol, 90+%
Thermo Scientific Chemicals
4-(4-Nitrophenylazo)resorcinol, 90+%
CAS: 74-39-5 | C12H9N3O4 | 259.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12232.1425 g
Catalog number ALFA12232.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4- (4-Nitrophenylazo)resorcinol
CAS74-39-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
View more
It is a derivitized resorcinol dye compound with an absorption maximum of 432 nm. Used as a pH indicator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a derivitized resorcinol dye compound with an absorption maximum of 432 nm. Used as a pH indicator.
Solubility
Soluble in dilute sodium hydroxide. Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Rosa Carballo; Alfonso Castiñeiras; Berta Covelo; Juan Niclós; Ezequiel M. Vázquez-López. Synthesis and characterization of a potassium complex of magneson: [K(HL)(OH2)2]∞ [H2L=4-(4-nitrophenylazo)resorcinol (magneson)]. Polyhedron, 2001, 20 (18), 2415-2420.
  2. Chuan Yang; Yuan Li; Cheng Huang. Determination of cationic surfactants in water samples by their enhanced resonance light scattering with azoviolet. Analytical and Bioanalytical Chemistry, 2002, 374, (5), 868-872.
  3. Reagent for Mg.