1-Dodecanol, 98%
1-Dodecanol, 98%
1-Dodecanol, 98%
Thermo Scientific Chemicals
1-Dodecanol, 98%
CAS: 112-53-8 | C12H26O | 186.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA12228.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Dodecanol
CAS112-53-8
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319
Health Hazard 3P264b-P280i-P305+P351+P338
View more
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Dodecanol is used as a cosmetic, textile auxiliaries, synthetic oil, emulsifiers and flotation agent of raw materials, a detergent raw material, a foaming agent of the toothpaste.
Solubility
Soluble in water (0.004g/L).
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
RUO – Research Use Only
General References:
  1. Rebecca A Hall; Kara J Turner; James Chaloupka; Fabien Cottier; Luisa De Sordi; Dominique Sanglard; Lonny R Levin; Jochen Buck; Fritz A Mühlschlegel. The quorum-sensing molecules farnesol/homoserine lactone and dodecanol operate via distinct modes of action in Candida albicans. Eukaryotic Cell, 2011, 10 (8), 1034-1042.
  2. Chris Grant; John M. Woodley; Frank Baganz; Chris Grant; John M. Woodley; Frank Baganz. Whole-cell bio-oxidation of n-dodecane using the alkane hydroxylase system ofP. putidaGPo1 expressed inE. coli. Enzyme and Microbial Technology, 2011, 48 (6), 480-486.