Maleic anhydride, 98+%
Maleic anhydride, 98+%
Maleic anhydride, 98+%
Maleic anhydride, 98+%
Thermo Scientific Chemicals
Maleic anhydride, 98+%
CAS: 108-31-6 | C4H2O3 | 98.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA12178.30
Price (MYR)
276.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
276.00
EA
Add to cart
Specifications
Chemical Name or MaterialMaleic anhydride
CAS108-31-6
Health Hazard 1H302-H314-H317-H334-H372-H500
Health Hazard 2GHS H Statement
H334-H314-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
View more
Maleic anhydride is used in the preparation of unsaturated polyester resins, 4-butanediol, fumairc acid, tetrahydrofuran and gamma-butyrolactone. It is also used as lubricating oil additives, dispersants, artificial sweeteners, flavor enhancements, floor polishes and corrosion inhibitors. It finds application in personal care products like hair sprays. Further, it is used in water treatment chemicals, detergents, pharmaceuticals and copolymers. It is an active substrate for Diels-Alder reactions and dimerizes to form cyclobutane tetracarboxylic dianhydride .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Maleic anhydride is used in the preparation of unsaturated polyester resins, 4-butanediol, fumairc acid, tetrahydrofuran and gamma-butyrolactone. It is also used as lubricating oil additives, dispersants, artificial sweeteners, flavor enhancements, floor polishes and corrosion inhibitors. It finds application in personal care products like hair sprays. Further, it is used in water treatment chemicals, detergents, pharmaceuticals and copolymers. It is an active substrate for Diels-Alder reactions and dimerizes to form cyclobutane tetracarboxylic dianhydride .
Solubility
Soluble in water, acetone and dioxane. Slightly soluble in methanol, chloroform and benzene.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases, strong reducing agents, alkali metals and amines.
RUO – Research Use Only
General References:
  1. Reactive dienophile. For a review of its Diels-Alder reactions, see: Org. React., 4, 1 (1948). For examples, see: Org. Synth. Coll., 3, 807 (1955); 4, 890 (1963). For reaction with Danishefsky's diene ( 1-Methoxy-3-trimethyl siloxy-1,3-butadiene, L14672 ), see: Org. Synth. Coll., 7, 312 (1990).
  2. Also undergoes photochemical and thermal (2+2)-cycloadditions with alkenes. For reaction with 1-alkynes, see: Chem. Ber., 102, 3974 (1969).
  3. For thermal (2+2)-cycloaddition with allene at high temperature, see: Org. Synth. Coll., 5, 459 (1973).
  4. For use in combination with Urea hydrogen peroxide adduct, L13940 , to generate peroxymaleic acid in epoxidation and Baeyer-Villiger reactions, see: Heterocycles, 36, 1075 (1993). Peroxymaleic acid can also be generated in situ from maleic anhydride and 30% H2O2 in NMP, and has been found to be a very effective oxidant for conversion of sulfides to sulfoxides. For a study of stoichiometric and catalytic oxidations with this system, see: J. Org. Chem., 61, 5693 (1996).
  5. Bao, Z.; Lee, E.; Tao, J.; Sun, X. Effect of halloysite and maleic anhydride grafted polypropylene on the isothermal crystallization kinetics of polypropylene based composites. Polym. Sci. Ser. A Polym. Phys. 2015, 57 (6), 889-897.
  6. Chatzidimitriou, A.; Bond, J. Q. Oxidation of levulinic acid for the production of maleic anhydride: breathing new life into biochemicals. Green Chem. 2015, 17 (8), 4367-4376.