1-Bromo-4-chlorobenzene, 98+%
1-Bromo-4-chlorobenzene, 98+%
1-Bromo-4-chlorobenzene, 98+%
1-Bromo-4-chlorobenzene, 98+%
Thermo Scientific Chemicals
1-Bromo-4-chlorobenzene, 98+%
CAS: 106-39-8 | C6H4BrCl | 191.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1000 g
Catalog number ALFA12160.30
Price (MYR)
329.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
329.00
EA
Add to cart
Specifications
Chemical Name or Material1-Bromo-4-chlorobenzene
Melting Point63°C to 67°C
CAS106-39-8
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-4-chlorobenzene is used as a pharmaceutical intermediate.
Solubility
Soluble in water 0.045g/L.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Harry A.J. Oonka; Peter R. van der Lindeb; Jan Huinink; Jacobus G. Blok. Representation and assessment of vapor pressure data; a novel approach applied to crystalline 1-bromo-4-chlorobenzene, 1-chloro-4-iodobenzene, and 1-bromo-4-iodobenzene. The Journal of Chemical Thermodynamics. 1998, 30 (7), 897-907.
  2. Vincenzo Calò; Angelo Nacci; Antonio Monopoli Dr.; and Pietro Cotugno. Heck Reactions with Palladium Nanoparticles in Ionic Liquids: Coupling of Aryl Chlorides with Deactivated Olefins. Angewandte Chemie. 2009, 121 (33), 6217-6219.