Succinic acid, 99+%
Succinic acid, 99+%
Succinic acid, 99+%
Thermo Scientific Chemicals
Succinic acid, 99+%
CAS: 110-15-6 | C4H6O4 | 118.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12084.36500 g
Catalog number ALFA12084.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSuccinic acid
CAS110-15-6
Melting Point186°C to 188°C
Boiling Point235°C
Health Hazard 1H318-H335
View more
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Succinic acid acts as a leaching agent in extracting lithium, cobalt from used Li-ion batteries and magnesium from magnesite ore. It is used as a starting material in the synthesis of new elastic polyesters, fumaric acid, succinic anhydride and alkyd resins. It plays an important role as an acidity regulator in the food and beverage industry, as an excipient in pharmaceutical products and acts as a cocrystallising agent in organic synthesis. Its derivative viz diethyl ester is used as a substrate in the Stobbe condensation.
Solubility
Soluble in ethanol, ethyl ether, acetone and methanol. Insoluble in toluene, benzene, carbon disulfide, carbon tetrachloride and petroleum ether.
Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only
General References:
  1. Hwang, H. J.; Choi, S. P.; Lee, S. Y.; Choi, J. I.; Han, S. J.; Lee, P. C. Dynamics of membrane fatty acid composition of succinic acid-producing Anaerobiospirillum succiniciproducens. J. Biotechnol. 2015, 193, 130-133.
  2. Gunnarsson, I. B.; Kuglarz, M.; Karakashev, D.; Angelidaki, I. Thermochemical pretreatments for enhancing succinic acid production from industrial hemp (Cannabis sativa L.). Bioresour. Technol. 2015, 182, 58-66.