Tetradecanoic acid, 98%
Tetradecanoic acid, 98%
Tetradecanoic acid, 98%
Tetradecanoic acid, 98%
Thermo Scientific Chemicals
Tetradecanoic acid, 98%
CAS: 544-63-8 | C14H28O2 | 228.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number ALFA12067.36
Price (MYR)
290.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
290.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetradecanoic acid
CAS544-63-8
Health Hazard 1H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280i-P305+P351+P338
View more
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a flow Agent and emulsifier. Used in artificial orris fragrances. Employed as anionic/nonionic Surfactants in soaps and detergents.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
RUO – Research Use Only
General References:
  1. Erika M Hernández.; Tze-Lee Phang.; Xinyun Wen.; Elias I Franses. Adsorption and direct probing of fibrinogen and sodium myristate at the air/water interface.J Colloid Interface Sci. 2002,250(2), 271-280.
  2. Wen X.; Franses EI. Effect of Protonation on the Solution and Phase Behavior of Aqueous Sodium Myristate.J Colloid Interface Sci. 2000,23142-51.