4-Ethylcatechol, 98%
4-Ethylcatechol, 98%
4-Ethylcatechol, 98%
4-Ethylcatechol, 98%
Thermo Scientific Chemicals
4-Ethylcatechol, 98%
CAS: 1124-39-6 | C8H10O2 | 138.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12048.065 g
Catalog number ALFA12048.06
Price (MYR)
2,246.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
2,246.00
EA
Add to cart
Specifications
Chemical Name or Material4-Ethylcatechol
CAS1124-39-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c