Pyridoxine hydrochloride, 99%
Pyridoxine hydrochloride, 99%
Pyridoxine hydrochloride, 99%
Pyridoxine hydrochloride, 99%
Thermo Scientific Chemicals
Pyridoxine hydrochloride, 99%
CAS: 58-56-0 | C8H12ClNO3 | 205.638 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALFA12041.36
Price (MYR)
1,874.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,874.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyridoxine hydrochloride
CAS58-56-0
Melting Point∼206°C (decomposition)
Recommended StorageAmbient temperatures
RTECS NumberUV1350000
View more
Pyridoxol hydrochlroride is a salt of pyridoxine free base which is used to treat low levels of vitamin B6. It is essential to maintain the health of nerves, skin and red blood cells. It is also used to treat nerve disorder and hereditary disorders like xanthurenic aciduria, cystathioninuria, hyperoxaluria and homocystinuria. It finds application in cosmetics, food and feed additives. It plays an important role in transporting protein Bsu1 and thiamine uptake. Furthermore, it is also involved in various biochemical reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Pyridoxol hydrochlroride is a salt of pyridoxine free base which is used to treat low levels of vitamin B6. It is essential to maintain the health of nerves, skin and red blood cells. It is also used to treat nerve disorder and hereditary disorders like xanthurenic aciduria, cystathioninuria, hyperoxaluria and homocystinuria. It finds application in cosmetics, food and feed additives. It plays an important role in transporting protein Bsu1 and thiamine uptake. Furthermore, it is also involved in various biochemical reactions.
Solubility
Soluble in water and alcohol. Slightly soluble in acetone. In soluble in ether and chloroform.
Notes
Light sensitive. Incompatible with bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Brahman, D.; Sinha, B. Non-covalent interactions between {N, N'-bis [(2-pyridinyl) methylene]-1, 2-benzenediamine]-bis (nitrato)} Cu(II) with pyridoxine hydrochloride in methanol at T=(298.15, 308.15 and 318.15) K. J. Chem. Thermodyn. 2014, 75, 136-144.
  2. Banipal, T. S.; Singh, H.; Banipal, P. K.; Singh, V. Volumetric and viscometric studies on L-ascorbic acid, nicotinic acid, thiamine hydrochloride and pyridoxine hydrochloride in water at temperatures (288.15-318.15) K and at atmospheric pressure. Thermochim. Acta 2013, 553, 31-39.