1-Octadecanol, 97%
1-Octadecanol, 97%
1-Octadecanol, 97%
Thermo Scientific Chemicals
1-Octadecanol, 97%
CAS: 112-92-5 | C18H38O | 270.501 g/mol
Have Questions?
Catalog NumberQuantity
ALFA12020.30250 g
Catalog number ALFA12020.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Octadecanol
CAS112-92-5
Melting Point56°C to 60°C
Recommended StorageAmbient temperatures
RTECS NumberRG2010000
View more
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.
Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.
Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
RUO – Research Use Only
General References:
  1. Thomas PS, et al. Electrical properties of natural rubber nanocomposites: effect of 1-octadecanol functionalization of carbon nanotubes.J. Mater. Sci.2012,47(7), 3344-49.
  2. Diana N H Tran.; Emma L Prime.; Michael Plazzer.; Andy H M Leung.; George Yiapanis.; Andrew J Christofferson.; Irene Yarovsky.; Greg G Qiao.; David H Solomon. Molecular interactions behind the synergistic effect in mixed monolayers of 1-octadecanol and ethylene glycol monooctadecyl ether.J. Phys. Chem. B,2013,117(13), 3603-3612.