Starch, from potato, soluble
Starch, from potato, soluble
Starch, from potato, soluble
Starch, from potato, soluble
Thermo Scientific Chemicals
Starch, from potato, soluble
CAS: 9005-84-9 | C12H22O11 | 342.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number ALFA11961.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialStarch
Name NoteFrom potato
CAS9005-84-9
Melting Point256°C to 258°C (decomposition)
Recommended StorageAmbient temperatures
View more
In food industry, starch is used as a food preservative. Starch has extensive industrial use as an adhesive or bonding agent. It is also used in paper industry. Starch is used in the procedure to manufacture gypsum wallboards. Starch used in the laundry, also known as clothing starch. Glucose derived from starch may be fermented further to make ethanol or biofuel. Starch is also employed in the petroleum exploration industry. It is interesting to note that starch may be used to make hydrogen by making usage of enzymes.